Learn More
Enzo Life Sciences Proteasome 20S alpha1, 2, 3, 5, 6 & 7 subunits monoclonal antibody (MCP231) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81950100
Encompass_Preferred
Clone: MCP231. Isotype: IgG1κ. Immunogen: Dinitrophenylated proteasomes. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Yeast, Rabbit, Potato. Specificity: Recognizes the α1, 2, 3, 5, 6 & 7 subunits of the 20S proteasome. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1593