Learn More
Enzo Life Sciences Proteasome 20S alpha4 subunit monoclonal antibody (MCP79) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW91400100
Encompass_Preferred
Clone: MCP79. Isotype: IgG1. Immunogen: Human placenta derived proteasomes. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Specificity: Recognizes the α4 subunit of the 20S proteasome. Applications: IHC, WB. Formulation: Liquid. Ascites in TBS, containing 5 mM sodium azide.
Catalog No. 50-201-1667