Learn More
Enzo Life Sciences Proteasome 20S alpha5 subunit (human) monoclonal antibody (HP903) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW82700100
Encompass_Preferred
Clone: HP903. Isotype: IgG. Immunogen: Human erythrocyte-derived proteasomes. Species reactivity: Human. Specificity: Recognizes the α5 subunit of the 20S proteasome. Applications: IHC, IP. Purity: Partially purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1613