Learn More
Enzo Life Sciences Proteasome 20S alpha6 subunit monoclonal antibody (MCP106) (1 ml)
SDP
Supplier:  Enzo Life Sciences BMLPW93900001
Encompass_Preferred
Clone: MCP106. Isotype: IgG1. Immunogen: Human placenta-derived proteasomes. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse Rabbit. Applications: WB. Formulation: Liquid. Tissue culture supernatant containing 10mM sodium azide.
Catalog No. 50-201-1684