Learn More
Enzo Life Sciences Proteasome 20S beta7 subunit (human) monoclonal antibody (MCP444) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW91500025
Encompass_Preferred
Clone: MCP444. Isotype: IgG1. Immunogen: Human placenta derived proteasomes. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes the β7 subunit of the 20S proteasome. Applications: IP, WB. Purity: Purified. Formulation: Liquid. Ascites in PBS, containing 0.01 M sodium azide.
Catalog No. 50-201-1668