Learn More
Enzo Life Sciences Proteasome 20S beta5i subunit monoclonal antibody (LMP7-1) (25 µl)
SDP
Catalog No. 502011647
Encompass_Preferred
Clone: LMP7-1. Isotype: IgG1. Immunogen: Recombinant human proteasome subunit β5i. Source: Purified from ascites. Species reactivity: Human, Rat. Specificity: Recognizes the β5i subunit of proteasome 20S. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 10mM sodium azide.
Catalog No. 50-201-1647 Supplier Enzo Life Sciences Supplier No. BMLPW88450025
Clone: LMP7-1. Isotype: IgG1. Immunogen: Recombinant human proteasome subunit β5i. Source: Purified from ascites. Species reactivity: Human, Rat. Specificity: Recognizes the β5i subunit of proteasome 20S. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 10mM sodium azide.