Learn More
Enzo Life Sciences Proteasome 20S alpha2 subunit monoclonal antibody (MCP21) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81050100
Encompass_Preferred
Clone: MCP21. Isotype: IgG1. Immunogen: Human placenta derived proteasomes. Source: Purified from mouse ascites. Species reactivity: Human Rabbit, Bovine. Specificity: Recognizes the α2 subunit of the 20S proteasome. Applications: IHC, IP, WB. Purity: Partially purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1570