Learn More
Enzo Life Sciences Proteasome 20S alpha5 subunit monoclonal antibody (MCP196) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81250100
Encompass_Preferred
Clone: MCP196. Isotype: IgG2a. Immunogen: Dinitrophenylated human placenta derived proteasomes. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Rabbit. Specificity: Recognizes the α5 subunit of the 20S proteasome. Applications: IHC, WB. Purity: Partially purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1577