Learn More
Enzo Life Sciences Proteasome 20S beta1 subunit monoclonal antibody (MCP421) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81400025
Encompass_Preferred
Clone: MCP421. Isotype: IgG1. Immunogen: Dinitrophenylated proteasomes. Source: Purified from mouse ascites. Species reactivity: Human Yeast, Rabbit. Specificity: Recognizes the β1 subunit of the 20S proteasome. Applications: IHC, WB. Purity: Protein G-affinity purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1582