Learn More
Enzo Life Sciences Proteasome 20S beta2 subunit monoclonal antibody (MCP165) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW93000025
Encompass_Preferred
Clone: MCP165. Isotype: IgG1. Immunogen: Dinitrophenylated proteasomes. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Specificity: Recognizes the β2 subunit of the 20S proteasome. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-1677