Learn More
Enzo Life Sciences Proteasome 20S beta5t subunit (human), monoclonal antibody (b8-17) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW10200100
Encompass_Preferred
Clone: b8-17. Isotype: IgG1. Immunogen: Recombinant human 20S proteasome β5t subunit. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Specificity: Recognizes the β5t subunit of the 20S proteasome. Applications: WB. Purity: Ion exchange purified. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1560