Learn More
Enzo Life Sciences Proteasome activator 11S beta subunit polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82400025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 15-31 of mouse PA28β. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes the β subunit of proteasome activator 11S. Applications: IHC, WB. Formulation: Liquid. Antiserum containing 10mM sodium azide.
Catalog No. 50-201-1603