Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail(EDTA-Free,100X in DMSO) 100x1ml
SDP
Catalog No. 5000064171
Encompass_Preferred
Protease inhibitor cocktails are routinely employed in protein extraction protocols to limit protein degradation due to intrinsic cellular protease activity facilitating the preservation of native protein structure and biochemical properties Upon cell lysis or tissue homogenization endogenous proteases including cysteine proteases serine proteases acid proteases and aminopeptidases become activated and can degrade target proteins This ready-to-use cocktail formulation supplied in DMSO at a 100X concentration specifically inhibits proteases via a defined blend of inhibitors (AEBSF Aprotinin Bestatin E-64 Leupeptin and Pepstatin A) selected to broadly target proteolytic activity encountered in mammalian cell extracts or tissue lysates This EDTA-free protease inhibitor cocktail is particularly suitable for downstream applications sensitive to divalent cations protein phosphorylation analysis or enzyme assays
Catalog No. 50-000-64171 Supplier Apexbio Technology LLC Supplier No. K1007100
Protease inhibitor cocktails are routinely employed in protein extraction protocols to limit protein degradation due to intrinsic cellular protease activity facilitating the preservation of native protein structure and biochemical properties Upon cell lysis or tissue homogenization endogenous proteases including cysteine proteases serine proteases acid proteases and aminopeptidases become activated and can degrade target proteins This ready-to-use cocktail formulation supplied in DMSO at a 100X concentration specifically inhibits proteases via a defined blend of inhibitors (AEBSF Aprotinin Bestatin E-64 Leupeptin and Pepstatin A) selected to broadly target proteolytic activity encountered in mammalian cell extracts or tissue lysates This EDTA-free protease inhibitor cocktail is particularly suitable for downstream applications sensitive to divalent cations protein phosphorylation analysis or enzyme assays