Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protease 7/OmpT E.c 100ug
SDP

Catalog No. 5000183154
Encompass_Preferred

Protease 7 (OmpT) is a recombinant Escherichia coli outer-membrane protease supplied as a His-tagged, lyophilized protein for research applications. It preferentially cleaves substrates with paired basic residues and is used in proteolytic assays, substrate specificity studies, and protein processing workflows. The formulation and handling guidance support activity preservation during storage and use.

  • Recombinant Escherichia coli OmpT protease with His tag.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation in Tris-EDTA with trehalose for stability.
  • Approximate molecular weight 37.5 kDa and full amino acid sequence available.
  • Reconstitution recommended at ≥100 μg/mL in ddH2O; stable at 4°C for one week after reconstitution.
  • Store lyophilized at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.

Catalog No. 50-001-83154 Supplier Medchemexpress LLC Supplier No. HYP71483100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protease 7 (OmpT) is a recombinant Escherichia coli outer-membrane protease supplied as a His-tagged, lyophilized protein for research applications. It preferentially cleaves substrates with paired basic residues and is used in proteolytic assays, substrate specificity studies, and protein processing workflows. The formulation and handling guidance support activity preservation during storage and use.

  • Recombinant Escherichia coli OmpT protease with His tag.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation in Tris-EDTA with trehalose for stability.
  • Approximate molecular weight 37.5 kDa and full amino acid sequence available.
  • Reconstitution recommended at ≥100 μg/mL in ddH2O; stable at 4°C for one week after reconstitution.
  • Store lyophilized at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.