Learn More
Abcam Protease and Phosphatase Inhibitor Cocktail, 1KIT
Catalog No. 502589200
Encompass_Preferred
Protease and phosphatase inhibitor cocktail prevents both proteolytic and phosphatase activities. AEBSF free and mass spectroscopy compatible – water soluble. - Cited in over 40 publications- It is a part of the multiplex kits range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9200 Supplier Abcam Supplier No. AB2011191KIT
Protease and phosphatase inhibitor cocktail prevents both proteolytic and phosphatase activities. AEBSF free and mass spectroscopy compatible – water soluble. - Cited in over 40 publications- It is a part of the multiplex kits range.
The product is subject to the following: Abcam Restricted Use Statement