Learn More
Sigma Aldrich Fine Chemicals Biosciences Protease from Bacillus licheniformis, 9014-01-1, MFCD00132092, 250mg
Catalog No. NC1644380
Encompass
Type VIII, lyophilized powder, 7-15 units/mg solid, Synonym: Proteinase from Bacillus licheniformis, Subtilisin A. The enzyme is found to be stable at pH 8-10, retaining activity of up to 90% for 24 hours. It shows maximum activity at temperatures between 55-60 °C.
Catalog No. NC1644380 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P5380250MG
Type VIII, lyophilized powder, 7-15 units/mg solid, Synonym: Proteinase from Bacillus licheniformis, Subtilisin A. The enzyme is found to be stable at pH 8-10, retaining activity of up to 90% for 24 hours. It shows maximum activity at temperatures between 55-60 °C.