Learn More
Medchemexpress LLC HY-K0010 1 mL × 50 Medchemexpress, Protease Inhibitor Cocktail (EDTA-Free, 100X in DMSO) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYK00101 ML W 50
Encompass_Preferred
Medchemexpress, HY-K0010 1 mL × 50 Protease Inhibitor Cocktail (EDTA-Free, 100X in DMSO) CAS: Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3276