Learn More
Sigma Aldrich Fine Chemicals Biosciences Protease from Streptomyces griseus, 9036-06-0, MFCD00132092, 1 g
Catalog No. NC0688085
Encompass
Type XIV, ≥3.5 units/mg solid, powder, Synonym: Actinase E, Pronase E. Highly stable in pH range of 5.0 to 9.0, with peak activity at pH 8.8. Compatible with many DNA and RNA isolation buffers. Broad substrate specificity.
Catalog No. NC0688085 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P51471G
Type XIV, ≥3.5 units/mg solid, powder, Synonym: Actinase E, Pronase E. Highly stable in pH range of 5.0 to 9.0, with peak activity at pH 8.8. Compatible with many DNA and RNA isolation buffers. Broad substrate specificity.