Learn More
TARGETMOL CHEMICALS INC Protease Inhibitor Cocktail (EDTA-Free, 100× in DMSO)
SDP
Catalog No. NC1998014
Encompass_Preferred
TargetMol provides a wide array of customizable Protease Inhibitor Cocktails, each with varying mechanisms of actions, aimed at protecting your protein samples. They work by helping proteins avoid rapid degradation caused by endogenous proteases during the cell lysis and protein purification processes. The Protease Inhibitor Cocktail I contains six protease inhibitors with broad specificity, aimed at facilitating the inhibition of aspartyl, cysteine, serine, as well as several aminopeptidases.
Catalog No. NC1998014 Supplier TARGETMOL CHEMICALS INC Supplier No. C0001100ML
Add to Cart
Add to Cart
TargetMol provides a wide array of customizable Protease Inhibitor Cocktails, each with varying mechanisms of actions, aimed at protecting your protein samples. They work by helping proteins avoid rapid degradation caused by endogenous proteases during the cell lysis and protein purification processes. The Protease Inhibitor Cocktail I contains six protease inhibitors with broad specificity, aimed at facilitating the inhibition of aspartyl, cysteine, serine, as well as several aminopeptidases.