Learn More
AG Scientific Inc Protease Inhib Cocktail II
Catalog No. 502522614
Encompass_Preferred
"Protease Inhibitor Cocktail II is a specially formulated cocktail of five protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, serine, and metalloproteases, as well as, aminopeptidases. Recommended for use with bacterial cell extracts. Used as a component of diagnostic cartidges for COVID-19 rapid testing. 5 mL solution recommended for the inhibition of proteases extracted from 20 g of E. coli. When reconstituted, each vial contains: 20 mM AEBSF HCl 1.7 mM Bestatin 200 uM E-64 85 mM EDTA 2mM Pepstatin A Preparation: Reconstitute each vial with 1 mL DMSO (first) and 4 mL water to obtain a 5 mL stock solution. Please note: the solution may appear turbid."
Catalog No. 50-252-2614 Supplier AG Scientific Inc Supplier No. P1511SOLID
"Protease Inhibitor Cocktail II is a specially formulated cocktail of five protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, serine, and metalloproteases, as well as, aminopeptidases. Recommended for use with bacterial cell extracts. Used as a component of diagnostic cartidges for COVID-19 rapid testing. 5 mL solution recommended for the inhibition of proteases extracted from 20 g of E. coli. When reconstituted, each vial contains: 20 mM AEBSF HCl 1.7 mM Bestatin 200 uM E-64 85 mM EDTA 2mM Pepstatin A Preparation: Reconstitute each vial with 1 mL DMSO (first) and 4 mL water to obtain a 5 mL stock solution. Please note: the solution may appear turbid."