Learn More
AG Scientific Inc Protease Inhib Cocktail VIII
Catalog No. 502522601
Encompass_Preferred
"Protease Inhibitor Cocktail VIII is a specially formulated mixture of three protease inhibitors with selective specificity for the inhibition of cysteine proteases, including calpains, cathepsins and papain. Each vial contains: 1.56 mM ALLN 500uM Antipain 1.5 mM E-64 Preparation: Supplied as a liquid in 1 mL DMSO. Add to water, aqueous buffer or directly to extraction media. "
Catalog No. 50-252-2601 Supplier AG Scientific Inc Supplier No. P15481ML
"Protease Inhibitor Cocktail VIII is a specially formulated mixture of three protease inhibitors with selective specificity for the inhibition of cysteine proteases, including calpains, cathepsins and papain. Each vial contains: 1.56 mM ALLN 500uM Antipain 1.5 mM E-64 Preparation: Supplied as a liquid in 1 mL DMSO. Add to water, aqueous buffer or directly to extraction media. "