Learn More
AG Scientific Inc Protease Inhib III NoAnmlDMSO
Supplier:  AG Scientific Inc P1543VIAL
Encompass_Preferred
"Protease Inhibitor Cocktail III, Animal-Free, DMSO-Free is a specially formulated mixture of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine and serine proteases as well as aminopeptidases. For applications that require animal-free reagents and where the use of DMSO is not recommended. 1 vial is recommended for the inhibition of proteases extracted from 20 g of tissue. When reconstituted to 1 mL, each vial contains: 100 mM AEBSF HCl 80 μM Aprotinin, recombinant (animal-free) 5 mM Bestatin 1.5 mM E-64 2 mM Leupeptin hemisulfate 1 mM Pepstatin A"
Catalog No. 50-252-2606