Learn More
AG Scientific Inc Protease Inhib V No Anml EDTA
Catalog No. 502522604
Encompass_Preferred
"Protease Inhibitor Cocktail V, EDTA-Free, Animal-Free is a mixture of four protease inhibitors which has been optimized for the inhibition of serine, cysteine but not metalloproteases. For applications that require animal-free reagents. 1X stock solution contains: 500 μM AEBSF HCl 150 nM Aprotinin, recombinant (animal-free) 1μM E-64 1μM Leupeptin hemisulfate Preparation: Reconstitute each vial with 1 mL water to obtain a 100X concentrated stock solution. Dilute with 100 mL solution to achieve 1X stock solution. Soluble in water, aqueous buffers, or add directly to extraction media. "
Catalog No. 50-252-2604 Supplier AG Scientific Inc Supplier No. P1544VIAL
"Protease Inhibitor Cocktail V, EDTA-Free, Animal-Free is a mixture of four protease inhibitors which has been optimized for the inhibition of serine, cysteine but not metalloproteases. For applications that require animal-free reagents. 1X stock solution contains: 500 μM AEBSF HCl 150 nM Aprotinin, recombinant (animal-free) 1μM E-64 1μM Leupeptin hemisulfate Preparation: Reconstitute each vial with 1 mL water to obtain a 100X concentrated stock solution. Dilute with 100 mL solution to achieve 1X stock solution. Soluble in water, aqueous buffers, or add directly to extraction media. "