Learn More
Active Motif Protease Inhibitor Cocktail
SDP
Catalog No. NC1847599
Encompass
Active Motif's Protease Inhibitor Cocktail is designed to prevent proteolytic degradation during lysis and extraction procedures from mammalian cells or tissues. The cocktail contains a broad-spectrum of inhibitors targeting cysteine proteases, serine proteases, aspartic acid proteases, and aminopeptidases. In addition to being ready-to-use, the Protease Inhibitor cocktail is also EDTA-free. The cocktail is supplied as a 100X stock solution in a liquid format that has been stabilized in dimethylsulfoxide (DMSO) for improved accuracy, solubility, and ease of use in comparison to traditional tablets.
Catalog No. NC1847599 Supplier Active Motif Supplier No. 37491
Add to Cart
Add to Cart
Active Motif's Protease Inhibitor Cocktail is designed to prevent proteolytic degradation during lysis and extraction procedures from mammalian cells or tissues. The cocktail contains a broad-spectrum of inhibitors targeting cysteine proteases, serine proteases, aspartic acid proteases, and aminopeptidases. In addition to being ready-to-use, the Protease Inhibitor cocktail is also EDTA-free. The cocktail is supplied as a 100X stock solution in a liquid format that has been stabilized in dimethylsulfoxide (DMSO) for improved accuracy, solubility, and ease of use in comparison to traditional tablets.