Learn More
Sigma Aldrich Fine Chemicals Biosciences Protease Inhibitor Cocktai
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I39111BO
Encompass
This protease inhibitor cocktail has been optimized and tested for mammalian cell and tissue extracts. It contains inhibitors with a broad specificity for serine cysteine and acid-proteases and aminopeptidases.
Catalog No. 50-290-1737