Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free, 100X in DMSO), 1mlx10
SDP
Catalog No. 501905545
Encompass_Preferred
Equivalent to Sigma P9599, for use with plant cell and tissue extracts to increase protein stability. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5545 Supplier Apexbio Technology LLC Supplier No. K101110
Equivalent to Sigma P9599, for use with plant cell and tissue extracts to increase protein stability. Other sizes are available. Please inqury us for quote.