Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail(EDTA-Free,100X in DMSO), 1ml x 50
SDP
Catalog No. 501905534
Encompass_Preferred
Equivalent to Sigma P8340, for use with cell and tissue extracts to increase protein stability Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5534 Supplier Apexbio Technology LLC Supplier No. K100750
Equivalent to Sigma P8340, for use with cell and tissue extracts to increase protein stability Other sizes are available. Please inqury us for quote.