Learn More
Bioworld Protease inhibitor Cocktail V Animal Free EDTA-Free, 1 Vial(s)
Catalog No. 50252993
Encompass_Preferred
Lyophillized blend of four protease inhibitors for use in applications that require animal free reagents. Reconstitute with lml water to produce a 100X concentrated solution.* Contains: AEBSF, Aprotinin (recombinant), E-64, Leupeptin,* Specificity: Serine Proteases, Cysteine Proteases, and Trypsin like ProteasesUsually ships within 24 hours.This product is sold for research use only.
Catalog No. 50-252-993 Supplier Bioworld Supplier No. 220200111
Add to Cart
Add to Cart
Lyophillized blend of four protease inhibitors for use in applications that require animal free reagents. Reconstitute with lml water to produce a 100X concentrated solution.* Contains: AEBSF, Aprotinin (recombinant), E-64, Leupeptin,* Specificity: Serine Proteases, Cysteine Proteases, and Trypsin like ProteasesUsually ships within 24 hours.This product is sold for research use only.