Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Protease Inhibitor Cocktail VI, Plant Cell, 1 Vial
SDP

Catalog No. 501977934
Encompass_Preferred

Mixture of six protease inhibitors for use with plant cell extracts. Supplied in 1ml DMSO. Will inhibit proteases extracted from 30g plant tissue.

Contains:

AEBSF
Bestatin
E-64
Leupeptin
Pepstatin A
Phenanthroline

Specificity:
Serine Proteases
Aminopeptidases
Cysteine
Proteases
Aspartic Proteases and Metalloproteases

Catalog No. 50-197-7934 Supplier Research Products International Corp Supplier No. P512001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mixture of six protease inhibitors for use with plant cell extracts. Supplied in 1ml DMSO. Will inhibit proteases extracted from 30g plant tissue.

Contains:

AEBSF
Bestatin
E-64
Leupeptin
Pepstatin A
Phenanthroline

Specificity:
Serine Proteases
Aminopeptidases
Cysteine
Proteases
Aspartic Proteases and Metalloproteases

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.