Learn More
Abcam Protease Inhibitor Cocktail, 1KIT
Catalog No. 502215267
Encompass_Preferred
Protease inhibitor cocktail is a water-soluble mixture used to inhibit a wide range of proteases during protein extraction, preventing protein degradation. - Cited in over 25 publications- It is a part of the multiplex kits range- Can be used with a variety of sample types, including mammalian tissue and cell extracts.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5267 Supplier Abcam Supplier No. AB656211KIT
Protease inhibitor cocktail is a water-soluble mixture used to inhibit a wide range of proteases during protein extraction, preventing protein degradation. - Cited in over 25 publications- It is a part of the multiplex kits range- Can be used with a variety of sample types, including mammalian tissue and cell extracts.
The product is subject to the following: Abcam Restricted Use Statement