Learn More
Bioworld Protease inhibitor Cocktail I-b, 1 Vial(s)
Catalog No. 50252986
Encompass_Preferred
Lyophilized blend of six inhibitors for inhibiting a broad range of proteases and esterases. 100x Solution* Contains: AEBSF, E-64, Leupeptin, Aprotinin, EDTA, Bestatin* Specificity: Serine Proteases, Esteroses, Cysteine Proteases, Metalloproteases and Trypsin like ProteasesThis product is sold for laboratory research use only.
Catalog No. 50-252-986 Supplier Bioworld Supplier No. 220200051
Lyophilized blend of six inhibitors for inhibiting a broad range of proteases and esterases. 100x Solution* Contains: AEBSF, E-64, Leupeptin, Aprotinin, EDTA, Bestatin* Specificity: Serine Proteases, Esteroses, Cysteine Proteases, Metalloproteases and Trypsin like ProteasesThis product is sold for laboratory research use only.