Learn More
Bioworld Protease Inhibitor Cocktail III (Animal Free for Mammalian Cells), 1 Vial(s)
Catalog No. 50252990
Encompass_Preferred
Mixture of six protease inhibitors for use in applications that require animal free reagents. Supplied in 1ml DMSO. Will treat proteases extracted from 20g tissue.Contains:100 mM AEBSF 80 μM Aprotinin, recombinant (animal free)5 mM Bestatin1.5 mME-642 mM LeupeptinHemisulfate1 mM Pepstatin ASpecificity: Serine Proteases, Cyteine Proteases Trypsin like Proteases, Aspartic Proteases as well as AminopeptidasesThis product is sold for research use only.
Catalog No. 50-252-990 Supplier Bioworld Supplier No. 220200081
Mixture of six protease inhibitors for use in applications that require animal free reagents. Supplied in 1ml DMSO. Will treat proteases extracted from 20g tissue.Contains:100 mM AEBSF 80 μM Aprotinin, recombinant (animal free)5 mM Bestatin1.5 mME-642 mM LeupeptinHemisulfate1 mM Pepstatin ASpecificity: Serine Proteases, Cyteine Proteases Trypsin like Proteases, Aspartic Proteases as well as AminopeptidasesThis product is sold for research use only.