Learn More
Bioworld Protease inhibitor Cocktail V EDTA Free, 1 Vial(s)
Catalog No. 50252994
Encompass_Preferred
Lyophilized blend of four protease inhibitors for inhibiting serine and cysteine but not metallo proteases. Reconstitute with 1ml of water to produce a l00X stock solution.* Contains: AEBSF, E-64, Aprotinin, Leupeptin* Specificity: Serine Proteases, Cysteine Proteases, and Trypsin like ProteasesUsually ships within 24 hours. This product is sold for research use only.
Catalog No. 50-252-994 Supplier Bioworld Supplier No. 220200121
Lyophilized blend of four protease inhibitors for inhibiting serine and cysteine but not metallo proteases. Reconstitute with 1ml of water to produce a l00X stock solution.* Contains: AEBSF, E-64, Aprotinin, Leupeptin* Specificity: Serine Proteases, Cysteine Proteases, and Trypsin like ProteasesUsually ships within 24 hours. This product is sold for research use only.