Learn More
Bioworld Protease inhibitor Cocktail VIII for Broad Range Cysteine Proteases, 1 mL
Catalog No. 50252997
Encompass_Preferred
Mixture of three protease inhibitors with specificity for inhibiting cysteine proteases. Supplied in lm DMSO.* Contains: ALLN, Antipain, E-64* Specificity: Calpain l/ll, Cathepsin A/B, Cathepsin L/S Papain, and Cysteine ProteasesUsuall ships within 24 hours.This product is sold for laboratory research use only.
Catalog No. 50-252-997 Supplier Bioworld Supplier No. 220200151
Mixture of three protease inhibitors with specificity for inhibiting cysteine proteases. Supplied in lm DMSO.* Contains: ALLN, Antipain, E-64* Specificity: Calpain l/ll, Cathepsin A/B, Cathepsin L/S Papain, and Cysteine ProteasesUsuall ships within 24 hours.This product is sold for laboratory research use only.