Learn More
Bioworld Protease inhibitor Cocktail VI for Plant Cell Extracts, 1 mL
Catalog No. 50252995
Encompass_Preferred
Mixture of six protease inhibitors for use with plant cell extracts. Supplied in lml DMSO. Will inhibit proteases extracted from 30g plant tissue.* Contains: AEBSF, Bestatin, E-64, Leupeptin, Pepstatin A, Phenanthroline* Specificity: Serine Proteases, Aminopeptidases, Cysteine Proteases, Aspartic Proteases and MetalloproteasesUsually ships within 24 hours.This product is sold for laboratory research use only.
Catalog No. 50-252-995 Supplier Bioworld Supplier No. 220200131
Mixture of six protease inhibitors for use with plant cell extracts. Supplied in lml DMSO. Will inhibit proteases extracted from 30g plant tissue.* Contains: AEBSF, Bestatin, E-64, Leupeptin, Pepstatin A, Phenanthroline* Specificity: Serine Proteases, Aminopeptidases, Cysteine Proteases, Aspartic Proteases and MetalloproteasesUsually ships within 24 hours.This product is sold for laboratory research use only.