Learn More
Bioworld Protease Inhibitor Cocktail III (for Mammalian Cells), 1 mL
Catalog No. 50252991
Encompass_Preferred
Mixture of six protease inhibitors for use with mammalian cells and tissue extracts. Supplied in l ml DMSO. Will treat proteases extracted from 20g tissue.* Contains: AEBSF, E-64, Aprotinin, Leupeptin, Bestatin, Pepstatin A* Specificity: Serine Proteases, Cysteine Proteases, Trypsin like Proteases, Aspartic Proteases, AminopeptidasesUsually ships within 24 hours.This product is sold for laboratory research use only.
Catalog No. 50-252-991 Supplier Bioworld Supplier No. 220200091
Mixture of six protease inhibitors for use with mammalian cells and tissue extracts. Supplied in l ml DMSO. Will treat proteases extracted from 20g tissue.* Contains: AEBSF, E-64, Aprotinin, Leupeptin, Bestatin, Pepstatin A* Specificity: Serine Proteases, Cysteine Proteases, Trypsin like Proteases, Aspartic Proteases, AminopeptidasesUsually ships within 24 hours.This product is sold for laboratory research use only.