Learn More
Bioworld Protease inhibitor Cocktail IV for Fungal and Yeast Extracts, 1 mL
Catalog No. 50252992
Encompass_Preferred
Mixture of four protease inhibitors for use with fungal and yeast extracts. Supplied in l ml DMSO. Will inhibit proteases extracted from 20g yeast.* Contains: AEBSF ,Pepstatin A, E-64, Phenanthroline* Specificity: Serine Protease, Cysteine Proteases, Aspartic Proteases and MetalloproteasesUsually ships within 24 hours.This product is sold for research use only.
Catalog No. 50-252-992 Supplier Bioworld Supplier No. 220200101
Mixture of four protease inhibitors for use with fungal and yeast extracts. Supplied in l ml DMSO. Will inhibit proteases extracted from 20g yeast.* Contains: AEBSF ,Pepstatin A, E-64, Phenanthroline* Specificity: Serine Protease, Cysteine Proteases, Aspartic Proteases and MetalloproteasesUsually ships within 24 hours.This product is sold for research use only.