Learn More
Bioworld Protease Inhibitor Cocktail II for Bacterial Cell Extracts, 1 Vial(s)
Catalog No. 50252989
Encompass_Preferred
Lyophilized mixture of five protease inhibitors for use with bacterial cell extracts. Reconstitute with 1ml DMSO and 4ml water to obtain a 5ml solution. Will treat proteases extracted from 20g E.Coli. * Contains: AEBSF, EDTA, Bestatin, E-64, Pepstatin A.* Specificity: Serine Proteases, Aminopeptidases, Cysteine Proteases, Metalloproteases and Aspartic Proteases.This product is sold for research use only.
Catalog No. 50-252-989 Supplier Bioworld Supplier No. 220200071
Add to Cart
Add to Cart
Lyophilized mixture of five protease inhibitors for use with bacterial cell extracts. Reconstitute with 1ml DMSO and 4ml water to obtain a 5ml solution. Will treat proteases extracted from 20g E.Coli. * Contains: AEBSF, EDTA, Bestatin, E-64, Pepstatin A.* Specificity: Serine Proteases, Aminopeptidases, Cysteine Proteases, Metalloproteases and Aspartic Proteases.This product is sold for research use only.