Learn More
Bioworld Protease inhibitor Cocktail VII for his-Tag®Sequences, 1 mL
Catalog No. 50252996
Encompass_Preferred
Mixture of five protease inhibitors for purification of proteins containing hexa histidine tagged sequences. Supplied in lml DMSO. Will inhibit proteases extracted from 10g of cells.* Contains: AEBSF, Bestatin, E-64, Pepstatin A, Phosphoromidon* Specificity: Serine Proteases, Cysteine Proteases, Aspartic Proteases, Aminopeptidoses and MetalloproteasesUsually ships within 24 hours.This product is sold for research use only
Catalog No. 50-252-996 Supplier Bioworld Supplier No. 220200141
Mixture of five protease inhibitors for purification of proteins containing hexa histidine tagged sequences. Supplied in lml DMSO. Will inhibit proteases extracted from 10g of cells.* Contains: AEBSF, Bestatin, E-64, Pepstatin A, Phosphoromidon* Specificity: Serine Proteases, Cysteine Proteases, Aspartic Proteases, Aminopeptidoses and MetalloproteasesUsually ships within 24 hours.This product is sold for research use only