Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free, 100X in DMSO), 1ml
SDP
Catalog No. 501905544
Encompass_Preferred
Equivalent to Sigma P9599, for use with plant cell and tissue extracts to increase protein stability. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5544 Supplier Apexbio Technology LLC Supplier No. K10111
Equivalent to Sigma P9599, for use with plant cell and tissue extracts to increase protein stability. Other sizes are available. Please inqury us for quote.