Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free, 100X in DMSO), 1ml x 10
SDP
Catalog No. 501905539
Encompass_Preferred
Equivalent to Sigma P8215, for use with fungal and yeast extracts to increase protein stability. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5539 Supplier Apexbio Technology LLC Supplier No. K100910
 Restricted Item
Add to Cart
Add to Cart
Equivalent to Sigma P8215, for use with fungal and yeast extracts to increase protein stability. Other sizes are available. Please inqury us for quote.