Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free, 100X in DMSO), 1mlx10
SDP
Supplier:  Apexbio Technology LLC K101010
Encompass_Preferred
Equivalent to Sigma P8849, for use in purification of Histidine-tagged proteins to increase protein stability. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5542
 Restricted Item