Learn More
Sigma Aldrich Fine Chemicals Biosciences Protease Inhibitor Cocktail, 1 mL
Catalog No. NC0530954
Encompass
After 48 hours in medium, add fresh inhibitors in fresh medium.
Catalog No. NC0530954 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P18601ML
After 48 hours in medium, add fresh inhibitors in fresh medium.