Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Protease Inhibitor Cocktail I, for General Use, Dry Powder, 1 Vial
SDP

Catalog No. 50488954
Encompass_Preferred

Lyophilized blend of five inhibitors for inhibiting a broad range of proteasesand esterases. Reconstitutes with 1 ml of water to produce a 100X concentrate solution.

Contains:

AEBSF
E-64
Leupeptin
Aprotinin
EDTA

Specificity:
Serine Proteases
Esterases
Cysteine
Proteases
Metalloproteases and Trypsin like Proteases

Catalog No. 50-488-954 Supplier Research Products International Corp Supplier No. P506001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lyophilized blend of five inhibitors for inhibiting a broad range of proteasesand esterases. Reconstitutes with 1 ml of water to produce a 100X concentrate solution.

Contains:

AEBSF
E-64
Leupeptin
Aprotinin
EDTA

Specificity:
Serine Proteases
Esterases
Cysteine
Proteases
Metalloproteases and Trypsin like Proteases

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.