Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Protease Inhibitor Cocktail II

Catalog No. AAJ64963LQ
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64963LQ 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64963LQ Supplier Thermo Scientific Chemicals Supplier No. J64963LQ
Add to Cart

 

Inhibits aspartic, cysteine, serine, and metalloproteases, as well as aminopeptidases. Recommended for use with bacterial cell extracts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits aspartic, cysteine, serine, and metalloproteases, as well as aminopeptidases
  • Recommended for use with bacterial cell extracts
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Protease Inhibitor Cocktail
Sensitivity Air sensitive
Content And Storage -30°C to -10°C
Form Lyophilized
For Use With (Application) Inhibits aspartic, cysteine, serine, and metalloproteases, as well as aminopeptidases. Recommended for use with bacterial cell extracts
Quantity 1 Unit
How much of the Protease Inhibitor Cocktail II should I use to inhibit protease activity?

Protease Inhibitor Cocktail II is recommended for use with bacterial cell extracts. We recommend a 5 mL stock solution of Protease Inhibitor Cocktail II for the inhibition of proteases extracted from 20 g of a bacterial pellet.

What is the composition of the Protease Inhibitor Cocktail II after reconstitution?

When reconstituted, each vial of Protease Inhibitor Cocktail II contains the following components:

  • 20 mM AEBSF.HCl
  • 1.7 mM Bestatin
  • 200 µM E-64
  • 85 mM EDTA
  • 2 mM Pepstatin A

How can I reconstitute Protease Inhibitor Cocktail II?

Protease Inhibitor Cocktail II is recommended for use with bacterial cell extracts. You should first reconstitute each vial in 1 mL DMSO. Then add 4 mL DI water to obtain a 5 mL stock solution. The solution may appear turbid. The 5 mL stock is enough to inhibit protease activity extracted from a 20 g bacterial cell pellet.

How should I store reconstituted stock solutions of Protease Inhibitor Cocktail II?

After reconstitution, we recommend that you aliquot and freeze the solution at -20°C. Stock solutions are stable for at least 1 month at -20°C.

The Protease Inhibitor Cocktail II solution appears turbid after reconstitution. Is this normal and will the solution remain turbid or clear up?

If the solution appears turbid after reconstitution, the solution for this material will appear turbid throughout its usage. This is normal and the material will function without issue.

How do I prepare the Protease Inhibitor Cocktail II?

We recommend that you reconstitute each vial with 1 mL DMSO first, and then add 4 mL water to obtain a 5 mL stock solution. The solution may appear turbid after reconstitution. 5 mL solution is recommended for the inhibition of proteases extracted from 20 g of E. coli. A 5 mL solution contains the following: 20 mM AEBSF.HCl, 1.7 mM bestatin, 200 µM E-64, 85 mM EDTA, and 2 mM pepstatin A.


RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.