Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Protease Inhibitor Cocktail IV, Fungal and Yeast Extracts, 1 Vial
SDP

Catalog No. 50488960
Encompass_Preferred

Mixture of four protease inhibitors for use with fungal and yeast extracts. Supplied in 1 ml DMSO. Will inhibit proteases extracted from 20g yeast.

Contains:

AEBSF
Pepstatin A
E-64
Phenanthroline

Specificity:
Serine Protease
Cysteine Proteases
Aspartic
Proteases and Metalloproteases

Catalog No. 50-488-960 Supplier Research Products International Corp Supplier No. P510001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mixture of four protease inhibitors for use with fungal and yeast extracts. Supplied in 1 ml DMSO. Will inhibit proteases extracted from 20g yeast.

Contains:

AEBSF
Pepstatin A
E-64
Phenanthroline

Specificity:
Serine Protease
Cysteine Proteases
Aspartic
Proteases and Metalloproteases

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.