Learn More
Research Products International Corp Protease Inhibitor Cocktail V, EDTA Free, 1 Vial
SDP
Catalog No. 50488958
Encompass_Preferred
Lyophilized blend of four protease inhibitors for inhibiting serine and cysteine but not metallo proteases. Reconstitute with 1ml of water to produce a l00X concentrated stock solution.
Contains:
AEBSF
E-64
Aprotinin
Leupeptin
Catalog No. 50-488-958 Supplier Research Products International Corp Supplier No. P509001
Lyophilized blend of four protease inhibitors for inhibiting serine and cysteine but not metallo proteases. Reconstitute with 1ml of water to produce a l00X concentrated stock solution.
Contains:
AEBSF
E-64
Aprotinin
Leupeptin