Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Protease Inhibitor Cocktails

Catalog No. pi78410
Change view
Click to view available options
Quantity:
1 mL
2 mL
Product Type:
Protease Inhibitor Cocktail
Protease Inhibitor Cocktail, EDTA-Free
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PI78410 2 mL Protease Inhibitor Cocktail
PI78415 1 mL Protease Inhibitor Cocktail, EDTA-Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI78410 Supplier Thermo Scientific™ Supplier No. 78410
Only null left
Add to Cart
Edge
Add to Cart

Protect your sample from proteolytic degradation during cell lysis with these broad-spectrum protease inhibitor cocktails.

Provide broad-spectrum protection against proteolytic degradation in cell lysis procedures with Thermo Scientific™ Protease Inhibitor Cocktails.

Both ready-to-use formulations inhibit most serine, cysteine, and calpain proteases that cause general degradation of proteins in cell extracts. Metalloproteases are inhibited as well. The EDTA-free formulation is ideal for preparing samples that will be analyzed by 2D gel electrophoresis. The refrigerator-stable, 100X liquid solutions are easy to use: simply add the appropriate volume of cocktail to the sample (10μL per 1mL of cell extract) and continue with your procedure. The Protease Inhibitor can be used with animal cells and tissues, bacteria, plants and yeast.

Highlights:

  • Ready-to-use, 100X liquid cocktail
  • Available with EDTA (supplied in separate vial) or as an EDTA-free formulation
  • Effective for suppressing proteolytic activity in detergent-based cell lysis reagents
  • No loss of valuable freezer space; cocktail is stable at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Formulation Proprietary components in DMSO
Product Type Protease Inhibitor Cocktail
Form Liquid
Sufficient For 200 mL of Sample
Quantity 2 mL
Composition Proprietary formulation
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.