Learn More
Abcam Protease Inhibitor Cocktail (100 x ), 3ML
Catalog No. 501963751
Encompass_Preferred
Protease inhibitor cocktail (100X) is a comprehensive reagent designed to inhibit proteases during protein extraction from various cell types, including mammalian, yeast, fungal cells, and bacteria. - Broad-spectrum inhibition – inhibits aminopeptidases, metalloproteases, and serine, cysteine, and aspartic acid proteases - Includes EDTA – chelates divalent metal ions for enhanced protection against metalloproteases - Compatible with most protein extraction – ideal for general-purpose protease inhibition - Form – 100X stock solutions of protease inhibitors and EDTA, supplied separately Note: For techniques incompatible with EDTA, use Protease Inhibitor Cocktail EDTA Free (ab270055)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-196-3751 Supplier Abcam Supplier No. AB2713063ML
Protease inhibitor cocktail (100X) is a comprehensive reagent designed to inhibit proteases during protein extraction from various cell types, including mammalian, yeast, fungal cells, and bacteria. - Broad-spectrum inhibition – inhibits aminopeptidases, metalloproteases, and serine, cysteine, and aspartic acid proteases - Includes EDTA – chelates divalent metal ions for enhanced protection against metalloproteases - Compatible with most protein extraction – ideal for general-purpose protease inhibition - Form – 100X stock solutions of protease inhibitors and EDTA, supplied separately Note: For techniques incompatible with EDTA, use Protease Inhibitor Cocktail EDTA Free (ab270055)
The product is subject to the following: Abcam Restricted Use Statement