Learn More
Research Products International Corp PROTEASE INHIBITOR COCKTAIL III,ANIMAL FREE,FOR MAMMALIAN CELLS, 1 VIAL
SDP
Catalog No. 50488956
Encompass_Preferred
A mixture of six protease inhibitors with animal free reagents for use with mammalian cells and tissue extracts. It is provided as a single vial. Solubilized in 1ml of DMSO.
Contains:
AEBSF HCl: 100 mM
Aprotinin (Recombinant): 80 uM
Bestatin: 5 mM
E-64: 1.5 mM
Leupeptin: 2 mM
Pepstatin: 1 mM
Serine Proteases
Cyteine Proteases
Trypsin like Proteases
Aspartic Proteases
Aminapeptidases
Catalog No. 50-488-956 Supplier Research Products International Corp Supplier No. P507501
A mixture of six protease inhibitors with animal free reagents for use with mammalian cells and tissue extracts. It is provided as a single vial. Solubilized in 1ml of DMSO.
Contains:
AEBSF HCl: 100 mM
Aprotinin (Recombinant): 80 uM
Bestatin: 5 mM
E-64: 1.5 mM
Leupeptin: 2 mM
Pepstatin: 1 mM
Serine Proteases
Cyteine Proteases
Trypsin like Proteases
Aspartic Proteases
Aminapeptidases